Assay Details
Gene Symbol: Smndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025400.3 | NP_001020571.1 | 6 | 1674 | 107 | |
NM_001035234.2 | NP_001030311.1 | 6 | 1836 | 107 | ||
NR_051995.1 | 6 | 1853 | 107 | |||
XM_006231608.2 | XP_006231670.1 | 6 | 2113 | 107 | ||
GenBank mRNA | CB726049.1 | - | 1 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
