Assay Details
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246483.1 | XP_006246545.1 | 1 - 2 | 39 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006246483.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
