Assay Details
Gene Symbol: Pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768472.1 | XP_008766694.1 | 9 | 1399 | 81 | |
XM_008768473.1 | XP_008766695.1 | 9 | 1333 | 81 | ||
XM_008768474.1 | XP_008766696.1 | 9 | 1323 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
