Assay Details
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768598.1 | XP_008766820.1 | 1 | 1107 | 89 | |
XM_008768599.1 | XP_008766821.1 | 1 | 1107 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768598.1, XM_008768599.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
