Assay Details
Gene Symbol: Nsun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768614.1 | XP_008766836.1 | 3 - 4 | 3192 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
