Assay Details
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031626.1 | NP_113814.1 | 10 | 1634 | 57 | |
GenBank mRNA | U14533.1 | - | 10 | 1634 | 57 | |
U20389.1 | - | 10 | 1578 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
