Assay Details
Gene Symbol: Bace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002802.1 | NP_001002802.1 | 2 - 3 | 395 | 70 | |
XM_006248153.2 | XP_006248215.1 | 2 - 3 | 449 | 70 | ||
GenBank mRNA | BC166597.1 | - | 2 - 3 | 409 | 7936295 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
