Assay Details
Gene Symbol: Vsig10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249437.2 | XP_006249499.1 | 1 | 1896 | 90 | |
XM_006249438.2 | XP_006249500.1 | 1 | 1896 | 90 | ||
XM_006249439.2 | XP_006249501.1 | 1 | 1896 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006249437.2, XM_006249438.2, XM_006249439.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
