Assay Details
Gene Symbol: Mmab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751201.3 | XP_003751249.1 | 2 - 3 | 219 | 78 | |
XM_008769355.1 | XP_008767577.1 | 2 - 3 | 219 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
