Assay Details
Gene Symbol: LOC100911204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234821.2 | XP_006234883.1 | 154 | 85 | ||
XM_006234822.2 | XP_006234884.1 | 384 | 85 | |||
XM_006234823.2 | XP_006234885.1 | 142 | 85 | |||
XM_008762127.1 | XP_008760349.1 | 202 | 85 | |||
XM_008762128.1 | XP_008760350.1 | 154 | 85 |
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170594.1 | NP_001164065.1 | 1 - 2 | 40 | 85 | |
XM_008762095.1 | XP_008760317.1 | 2 - 3 | 154 | 85 | ||
XM_008762096.1 | XP_008760318.1 | 2 - 3 | 384 | 85 | ||
XM_008762097.1 | XP_008760319.1 | 2 - 3 | 197 | 85 | ||
XM_008762098.1 | XP_008760320.1 | 2 - 3 | 154 | 85 | ||
XM_008762099.1 | XP_008760321.1 | 2 - 3 | 141 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
