Assay Details
Gene Symbol: LOC100911204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234821.2 | XP_006234883.1 | 486 | 75 | ||
XM_006234822.2 | XP_006234884.1 | 716 | 75 | |||
XM_006234823.2 | XP_006234885.1 | 474 | 75 | |||
XM_008762127.1 | XP_008760349.1 | 534 | 75 | |||
XM_008762128.1 | XP_008760350.1 | 399 | 75 |
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170594.1 | NP_001164065.1 | 6 - 7 | 300 | 75 | |
XM_008762095.1 | XP_008760317.1 | 9 - 10 | 486 | 75 | ||
XM_008762096.1 | XP_008760318.1 | 9 - 10 | 716 | 75 | ||
XM_008762097.1 | XP_008760319.1 | 9 - 10 | 529 | 75 | ||
XM_008762098.1 | XP_008760320.1 | 7 - 8 | 399 | 75 | ||
XM_008762099.1 | XP_008760321.1 | 9 - 10 | 473 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
