Assay Details
Gene Symbol: Sacs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252167.2 | XP_006252229.1 | 11 | 11941 | 94 | |
XM_006252168.2 | XP_006252230.1 | 10 | 11469 | 94 | ||
XM_006252169.2 | XP_006252231.1 | 10 | 11914 | 94 | ||
XM_008770792.1 | XP_008769014.1 | 10 | 11794 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
