Assay Details
Gene Symbol: Adra1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252107.2 | XP_006252169.1 | 2 | 456 | 96 | |
XM_006252108.2 | XP_006252170.1 | 2 | 454 | 96 | ||
XM_006252109.2 | XP_006252171.1 | 2 | 455 | 96 | ||
XM_006252110.2 | XP_006252172.1 | 2 | 450 | 96 | ||
XM_008770744.1 | XP_008768966.1 | 2 | 456 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006252107.2, XM_006252108.2, XM_006252109.2, XM_006252110.2, XM_008770744.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
