Assay Details
Gene Symbol: Crp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY310153.1 | - | 683 | 163 | ||
AY318961.1 | - | 683 | 163 | |||
AY321318.1 | - | 680 | 163 | |||
AY321324.1 | - | 683 | 163 | |||
AY325196.1 | - | 683 | 163 | |||
AY325218.1 | - | 683 | 163 | |||
AY325232.1 | - | 683 | 163 |
Gene Symbol: LOC501503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ100477.1 | - | 2 | 4296 | 163 |
Gene Symbol: Lrrgt00014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U83119.1 | - | 1 | 2599 | 163 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
