Assay Details
Gene Symbol: Rpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053971.3 | NP_446423.2 | 3 | 424 | 87 | |
XM_006249377.1 | XP_006249439.1 | 4 | 443 | 87 | ||
XM_006249378.2 | XP_006249440.1 | 4 | 477 | 87 | ||
GenBank mRNA | BC061784.1 | - | 3 | 420 | 87 | |
BC078761.1 | - | 3 | 427 | 7123086 | 87 | |
BC097295.1 | - | 3 | 428 | 7446680 | 87 | |
FQ209419.1 | - | 2 | 102 | 87 | ||
FQ213055.1 | - | 2 | 102 | 87 | ||
FQ213078.1 | - | 2 | 102 | 87 | ||
FQ216758.1 | - | 2 | 102 | 87 | ||
FQ217744.1 | - | 2 | 102 | 87 | ||
FQ219956.1 | - | 2 | 102 | 87 | ||
FQ219982.1 | - | 2 | 102 | 87 | ||
FQ220180.1 | - | 2 | 102 | 87 | ||
FQ220346.1 | - | 2 | 102 | 87 | ||
FQ220362.1 | - | 2 | 102 | 87 | ||
FQ220471.1 | - | 2 | 102 | 87 | ||
FQ220678.1 | - | 2 | 102 | 87 | ||
FQ220699.1 | - | 2 | 102 | 87 | ||
FQ220887.1 | - | 2 | 102 | 87 | ||
FQ220959.1 | - | 2 | 102 | 87 | ||
FQ221615.1 | - | 2 | 102 | 87 | ||
FQ222080.1 | - | 2 | 102 | 87 | ||
FQ222331.1 | - | 2 | 102 | 87 | ||
FQ222465.1 | - | 2 | 102 | 87 | ||
FQ222658.1 | - | 2 | 102 | 87 | ||
FQ223301.1 | - | 2 | 102 | 87 | ||
FQ226632.1 | - | 2 | 88 | |||
FQ227060.1 | - | 2 | 102 | 87 | ||
FQ227100.1 | - | 2 | 102 | 87 | ||
FQ227955.1 | - | 2 | 102 | 87 | ||
FQ227974.1 | - | 2 | 102 | 87 | ||
FQ228583.1 | - | 2 | 102 | 87 | ||
FQ228665.1 | - | 2 | 102 | 87 | ||
FQ229345.1 | - | 2 | 102 | 87 | ||
FQ229355.1 | - | 2 | 102 | 87 | ||
FQ229484.1 | - | 4 | 442 | 87 | ||
FQ229571.1 | - | 2 | 102 | 87 | ||
FQ229624.1 | - | 2 | 102 | 87 | ||
FQ229675.1 | - | 2 | 102 | 87 | ||
FQ229820.1 | - | 2 | 102 | 87 | ||
FQ229854.1 | - | 2 | 102 | 87 | ||
FQ229857.1 | - | 2 | 102 | 87 | ||
FQ229943.1 | - | 4 | 87 | |||
FQ230171.1 | - | 2 | 102 | 87 | ||
FQ230222.1 | - | 2 | 102 | 87 | ||
FQ232778.1 | - | 2 | 102 | 87 | ||
FQ233475.1 | - | 2 | 102 | 87 | ||
FQ234880.1 | - | 4 | 441 | 87 | ||
FQ235095.1 | - | 2 | 102 | 87 | ||
X87107.1 | - | 4 | 438 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
