Assay Details
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008891.2 | NP_001008891.2 | 8 | 1703 | 122 | |
XM_006253835.2 | XP_006253897.1 | 9 | 1755 | 123 | ||
GenBank mRNA | BC098008.1 | - | 8 | 1703 | 7454159 | 122 |
FQ230054.1 | - | 1 | 421 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
