Assay Details
Gene Symbol: LOC686142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751717.3 | XP_003751765.1 | 6 | 1108 | 75 | |
XM_006254133.2 | XP_006254195.1 | 5 | 1711 | 75 | ||
XM_006254135.2 | XP_006254197.1 | 5 | 852 | 75 |
Gene Symbol: LOC102553208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254077.2 | XP_006254139.1 | 5 | 1070 | 75 | |
XM_006254078.2 | XP_006254140.1 | 5 | 1223 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
