Assay Details
Gene Symbol: LOC686142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751717.3 | XP_003751765.1 | 2 - 3 | 387 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003751717.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
