Assay Details
Gene Symbol: F11r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053796.1 | NP_446248.1 | 5 | 545 | 100 | |
GenBank mRNA | AF241261.1 | - | 1 | 92 | 100 | |
AF276998.1 | - | 5 | 545 | 100 | ||
BC065309.1 | - | 5 | 527 | 5599898 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
