Assay Details
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC061847.1 | - | 1 | 43 | 5621065 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC061847.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
