Assay Details
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104630.1 | NP_001098100.1 | 9 | 1853 | 68 | |
XM_006254266.2 | XP_006254328.1 | 9 | 3480 | 68 | ||
GenBank mRNA | AY724491.1 | - | 9 | 1853 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
