Assay Details
Gene Symbol: Psmg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106138.2 | NP_001099608.2 | 7 | 115 | ||
GenBank mRNA | BM387084.1 | - | 1 | 120 | 115 | |
FQ212569.1 | - | 7 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
