Assay Details
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039004.1 | NP_001034093.1 | 1 | 100 | 92 | |
XM_006255132.2 | XP_006255194.1 | 1 | 733 | 92 | ||
GenBank mRNA | BC098748.1 | - | 1 | 100 | 7365565 | 92 |
BC168149.1 | - | 1 | 739 | 7457021 | 92 | |
FQ213930.1 | - | 1 | 143 | 92 | ||
FQ229498.1 | - | 1 | 141 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC098748.1, BC168149.1, NM_001039004.1, XM_006255132.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
