Assay Details
Gene Symbol: Slc7a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232323.2 | XP_006232385.1 | 1 | 281 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232323.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
