Assay Details
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031120.2 | NP_112382.2 | 2 | 2617 | 98 | |
GenBank mRNA | BC062015.1 | - | 2 | 2617 | 5621495 | 98 |
FQ216142.1 | - | 1 | 970 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
