Assay Details
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100533.1 | NP_001094003.1 | 6 | 1636 | 68 | |
XM_006233679.2 | XP_006233741.1 | 6 | 8735 | 68 | ||
XM_008761623.1 | XP_008759845.1 | 5 | 2839 | 68 | ||
GenBank mRNA | BC092608.1 | - | 5 | 1440 | 68 | |
FQ210633.1 | - | 6 | 1045 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
