Assay Details
Gene Symbol: Gtf3c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079941.1 | NP_001073410.1 | 12 | 2500 | 64 | |
XM_006233835.2 | XP_006233897.1 | 12 | 2491 | 64 | ||
GenBank mRNA | BC129109.1 | - | 12 | 2500 | 7105121 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
