Assay Details
Gene Symbol: Tgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019386.2 | NP_062259.2 | 13 | 2679 | 81 | |
GenBank mRNA | AF106325.1 | - | 12 | 2629 | 81 | |
BC062062.1 | - | 13 | 2679 | 5599202 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
