Assay Details
Gene Symbol: Cdh22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019161.2 | NP_062034.2 | 2 | 214 | 60 | |
GenBank mRNA | D83348.1 | - | 2 | 220 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D83348.1, NM_019161.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
