Assay Details
Gene Symbol: Zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127293.1 | NP_001120765.1 | 10 | 1698 | 75 | |
GenBank mRNA | BC166834.1 | - | 10 | 1698 | 9022075 | 75 |
FQ226567.1 | - | 5 | 976 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
