Assay Details
Gene Symbol: Smg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014243.1 | NP_001014265.1 | 14 | 1818 | 78 | |
XM_006228450.2 | XP_006228512.1 | 14 | 2749 | 78 | ||
GenBank mRNA | BC087051.1 | - | 14 | 1818 | 7115791 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
