Assay Details
Gene Symbol: Tmem43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007745.1 | NP_001007746.1 | 12 | 1175 | 82 | |
GenBank mRNA | BC083626.1 | - | 12 | 1175 | 7134239 | 82 |
FQ221746.1 | - | 13 | 1336 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
