Assay Details
Gene Symbol: U2af1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228776.2 | XP_006228838.1 | 8 | 777 | 66 | |
XM_006228777.1 | XP_006228839.1 | 7 | 661 | 66 | ||
XM_006228778.2 | XP_006228840.1 | 7 | 681 | 66 | ||
XM_006228779.2 | XP_006228841.1 | 7 | 678 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
