Assay Details
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF073379.1 | - | 885 | 86 | ||
L34938.1 | - | 412 | 86 | |||
U29873.1 | - | 885 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
