Assay Details
Gene Symbol: Htr1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764208.1 | XP_008762430.1 | 1 | 77 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764208.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
