Assay Details
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013059.1 | NP_037191.1 | 12 | 2301 | 120 | |
XM_006239136.2 | XP_006239198.1 | 12 | 2675 | 121 | ||
GenBank mRNA | BC088399.1 | - | 12 | 2324 | 7128677 | 121 |
J03572.1 | - | 12 | 2301 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
