Assay Details
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013170.2 | NP_001013188.2 | 11 | 1849 | 81 | |
GenBank mRNA | AY724504.1 | - | 11 | 1849 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
