Assay Details
Gene Symbol: Ift27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130495.1 | NP_001123967.1 | 7 | 897 | 95 | |
XM_006241931.2 | XP_006241993.1 | 7 | 906 | 95 | ||
GenBank mRNA | BC168150.1 | - | 907 | 7457249 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
