Assay Details
Gene Symbol: Arf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080904.2 | NP_543180.1 | 4 | 959 | 83 | |
XM_006257320.2 | XP_006257382.1 | 5 | 1168 | 83 | ||
GenBank mRNA | BC088865.1 | - | 4 | 959 | 7319260 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
