Assay Details
Gene Symbol: Serpine2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019197.1 | NP_062070.1 | 9 | 1621 | 86 | |
XM_006245161.2 | XP_006245223.1 | 9 | 2284 | 86 | ||
XM_006245162.2 | XP_006245224.1 | 10 | 1748 | 86 | ||
XM_008767212.1 | XP_008765434.1 | 9 | 1608 | 86 | ||
GenBank mRNA | BQ199747.1 | - | 1 | 86 | ||
CF110312.1 | - | 5 | 779 | 86 | ||
DV726078.1 | - | 3 | 501 | 86 | ||
FQ228974.1 | - | 7 | 1190 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
