Assay Details
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106282.1 | NP_001099752.1 | 8 | 866 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
