Assay Details
Gene Symbol: Aldh3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031731.2 | NP_113919.2 | 1 | 101 | 85 | |
XM_006246529.2 | XP_006246591.1 | 1 | 599 | 85 | ||
GenBank mRNA | FQ210485.1 | - | 1 | 164 | 85 | |
FQ221576.1 | - | 1 | 123 | 85 | ||
M73714.1 | - | 1 | 107 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M73714.1, NM_031731.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
