Assay Details
Gene Symbol: Acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034951.1 | NP_001030123.1 | 16 | 2753 | 89 | |
GenBank mRNA | BC099826.1 | - | 16 | 2753 | 7934093 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
