Assay Details
Gene Symbol: Bcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031752.2 | NP_113940.1 | 15 | 2140 | 90 | |
GenBank mRNA | BC072479.1 | - | 15 | 2140 | 7104047 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
