Assay Details
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021869.2 | NP_068641.2 | 10 | 1680 | 80 | |
XM_006227691.2 | XP_006227753.1 | 11 | 3002 | 80 | ||
XM_008758626.1 | XP_008756848.1 | 11 | 2996 | 80 | ||
GenBank mRNA | BC085737.1 | - | 10 | 1680 | 7193349 | 80 |
Gene Symbol: LOC100910446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758670.1 | XP_008756892.1 | 8 | 1566 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
