Assay Details
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007731.1 | NP_001007732.1 | 5 | 1880 | 111 | |
XM_006253320.2 | XP_006253382.1 | 5 | 1933 | 111 | ||
GenBank mRNA | BC078959.1 | - | 5 | 1880 | 7110849 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
