Assay Details
Gene Symbol: Snrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244057.2 | XP_006244119.1 | 7 | 3064 | 117 | |
XM_006244058.2 | XP_006244120.1 | 7 | 3098 | 117 | ||
XM_006244059.1 | XP_006244121.1 | 7 | 3057 | 117 | ||
XM_006244060.2 | XP_006244122.1 | 6 | 3072 | 117 | ||
XM_006244061.1 | XP_006244123.1 | 6 | 3078 | 117 | ||
XM_006244062.2 | XP_006244124.1 | 6 | 3006 | 117 | ||
XM_008766666.1 | XP_008764888.1 | 6 | 2997 | 117 | ||
XM_008766667.1 | XP_008764889.1 | 6 | 3139 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
