Assay Details
Gene Symbol: Senp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012355.1 | NP_001012355.1 | 2 - 3 | 62 | 80 | |
GenBank mRNA | BC089939.1 | - | 2 - 3 | 62 | 80 | |
FQ227853.1 | - | 2 - 3 | 75 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC089939.1, NM_001012355.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
