Assay Details
Gene Symbol: Rcan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012746.1 | NP_001012764.1 | 1 - 2 | 207 | 63 | |
GenBank mRNA | AY919305.1 | - | 1 - 2 | 207 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
