Assay Details
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013047.1 | NP_001013065.1 | 10 - 11 | 1093 | 58 | |
XM_008764483.1 | XP_008762705.1 | 9 - 10 | 1126 | 58 | ||
XM_008764484.1 | XP_008762706.1 | 9 - 10 | 1126 | 58 | ||
GenBank mRNA | BC079400.1 | - | 10 - 11 | 1240 | 58 | |
BC091225.1 | - | 10 - 11 | 1093 | 58 | ||
FQ224358.1 | - | 9 - 10 | 1131 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
